Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003279.2 | NP_003270.1 | 4 - 5 | 383 | 172 | |
XM_011529031.2 | XP_011527333.1 | 4 - 5 | 393 | 172 | ||
GenBank mRNA | AI080746.1 | - | 2 - 3 | 172 | ||
AK291323.1 | - | 4 - 5 | 381 | 172 | ||
BC005323.1 | - | 4 - 5 | 381 | 3934188 | 172 | |
BU854061.1 | - | 4 - 5 | 414 | 172 | ||
CR541864.1 | - | 3 - 4 | 317 | 172 | ||
CR541884.1 | - | 3 - 4 | 317 | 172 | ||
X07898.1 | - | 4 - 5 | 381 | 172 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |