Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136536.4 | NP_001130008.2 | 2 - 3 | 747 | 139 | |
NM_001170700.2 | NP_001164171.1 | 2 - 3 | 902 | 139 | ||
XM_006714014.3 | XP_006714077.1 | 3 - 4 | 1381 | 139 | ||
XM_011513693.2 | XP_011511995.1 | 3 - 4 | 1382 | 139 | ||
XM_011513695.2 | XP_011511997.1 | 3 - 4 | 1381 | 139 | ||
XM_011513696.2 | XP_011511998.1 | 2 - 3 | 765 | 139 | ||
XM_017008191.1 | XP_016863680.1 | 3 - 4 | 1382 | 139 | ||
XM_017008192.1 | XP_016863681.1 | 3 - 4 | 1384 | 139 | ||
XM_017008193.1 | XP_016863682.1 | 3 - 4 | 1383 | 139 | ||
GenBank mRNA | AK131492.1 | - | 2 - 3 | 740 | 139 | |
AK304357.1 | - | 2 - 3 | 733 | 139 | ||
AK316523.1 | - | 2 - 3 | 733 | 139 | ||
BC157885.1 | - | 2 - 3 | 902 | 9057096 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |