Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000429112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000429112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000429112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024520.hg.1 |
TC06001977.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024520.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024520.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024520.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024520.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024526.hg.1 |
TC06001977.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024526.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187669.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187669.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187669.hg.1 |
TC0600012709.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000429112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000429112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000429112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012709.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
ENST00000429112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
ENST00000429112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187662.hg.1 |
TC0600012709.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600187665.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187665.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187665.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187665.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187665.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187665.hg.1 |
ENST00000254765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187665.hg.1 |
ENST00000474760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187665.hg.1 |
ENST00000489134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187665.hg.1 |
NM_022361.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187665.hg.1 |
NR_024539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187665.hg.1 |
TC0600012709.hg.1 |
Clariom D TC |