Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007420.hg.1 |
TC01000478.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007420.hg.1 |
ENST00000373045.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007420.hg.1 |
ENST00000397631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007420.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007420.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007424.hg.1 |
TC01000478.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007424.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007424.hg.1 |
ENST00000373045.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007424.hg.1 |
ENST00000397631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007424.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007424.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007424.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007424.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014405.hg.1 |
ENST00000424794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155243.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155243.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155243.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155243.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155243.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155243.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155243.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155243.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155243.hg.1 |
TC0100007845.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100155244.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155244.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155244.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155244.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155244.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155244.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155244.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155244.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155244.hg.1 |
TC0100007845.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155250.hg.1 |
TC0100007845.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014405.hg.1 |
ENST00000424794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155245.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155245.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155245.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155245.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155245.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155245.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155245.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155245.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155245.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155245.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155245.hg.1 |
TC0100007845.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155249.hg.1 |
TC0100007845.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000329006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000373045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000397631.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000525897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
ENST00000532512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001031740.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_001113482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007845.hg.1 |
NM_152496.2 |
Refseq |