Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GANC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301409.1 | NP_001288338.1 | 5 - 6 | 1031 | 121 | |
NM_198141.2 | NP_937784.2 | 4 - 5 | 644 | 121 | ||
GenBank mRNA | AF545045.1 | - | 4 - 5 | 338 | 121 | |
AF545046.1 | - | 4 - 5 | 338 | 121 | ||
AK074037.1 | - | 4 - 5 | 536 | 121 | ||
AK074330.1 | - | 5 - 6 | 1033 | 121 | ||
BC059406.1 | - | 4 - 5 | 516 | 30330796 | 121 | |
BC093833.1 | - | 4 - 5 | 361 | 7939678 | 121 | |
BC112051.1 | - | 4 - 5 | 361 | 8327519 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |