Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010874.hg.1 |
TC02000704.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010874.hg.1 |
ENST00000331203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010874.hg.1 |
ENST00000409903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010874.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000272559.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
ENST00000472377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_001128165.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008904.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162169.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162169.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162169.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162169.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162169.hg.1 |
ENST00000331203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162169.hg.1 |
ENST00000409903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162169.hg.1 |
ENST00000441565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162169.hg.1 |
NM_153214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162169.hg.1 |
TC0200008904.hg.1 |
Clariom D TC |