Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000529349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000529349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000529349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000529349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013047.hg.1 |
ENST00000530218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013047.hg.1 |
ENST00000532404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013047.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013047.hg.1 |
TC15001356.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000529349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000529349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000529349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000529349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009393.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
ENST00000529349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
ENST00000529349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
ENST00000530218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
ENST00000532404.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
ENST00000559105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
ENST00000616326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
NM_203494.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169004.hg.1 |
TC1500009393.hg.1 |
Clariom D TC |