Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145030.1 | NP_001138502.1 | 13 - 14 | 4029 | 110 | |
XM_011533694.2 | XP_011531996.1 | 13 - 14 | 4029 | 110 | ||
XM_017006361.1 | XP_016861850.1 | 13 - 14 | 4029 | 110 | ||
XM_017006362.1 | XP_016861851.1 | 13 - 14 | 4029 | 110 | ||
GenBank mRNA | BX648094.1 | - | 12 - 13 | 2531 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons |