Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025160.hg.1 |
ENST00000245541.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025160.hg.1 |
ENST00000351904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025160.hg.1 |
ENST00000537584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025160.hg.1 |
ENST00000537686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025160.hg.1 |
NM_001172703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025160.hg.1 |
NM_001172704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025160.hg.1 |
NM_014001.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025160.hg.1 |
NM_138619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025160.hg.1 |
TC17001874.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025167.hg.1 |
ENST00000537584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025167.hg.1 |
NM_001172703.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025167.hg.1 |
NM_001172704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025167.hg.1 |
TC17001874.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
ENST00000578348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
ENST00000579743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
ENST00000582486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
ENST00000582717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
NM_001172703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
NM_001172704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
ENST00000578348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
ENST00000579743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
ENST00000582486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
ENST00000582717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
NM_001172703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
NM_001172704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195299.hg.1 |
TC1700011743.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700195302.hg.1 |
NM_001172703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195302.hg.1 |
NM_001172704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195302.hg.1 |
NM_001172703.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195302.hg.1 |
NM_001172704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195302.hg.1 |
TC1700011743.hg.1 |
Clariom D TC |