Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000378654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000433022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000433022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000433022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000433022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000433022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000433022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000434484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000434484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000434484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000434484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000434484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000434484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000434484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000434484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000434484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000467222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000467222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000467222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000467222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000467222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000467222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000467222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000467222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000470760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000470760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000470760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000470760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000470760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000470760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000481279.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000481279.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000481279.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000484941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000484941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000484941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000493152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000493152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000493152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000493152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000493152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000497202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000497202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000497202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000497202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000497202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000497202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000497202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
ENST00000497202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008873.hg.1 |
NM_003850.2 |
Refseq |