Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252043.1 | NP_001238972.1 | 3 - 4 | 1018 | 79 | |
NM_004783.3 | NP_004774.1 | 3 - 4 | 1018 | 79 | ||
NM_016151.3 | NP_057235.2 | 3 - 4 | 1018 | 79 | ||
XM_011545982.1 | XP_011544284.1 | 3 - 4 | 1244 | 79 | ||
XM_011545983.2 | XP_011544285.1 | 3 - 4 | 1244 | 79 | ||
XM_011545984.1 | XP_011544286.1 | 3 - 4 | 1244 | 79 | ||
XM_011545985.1 | XP_011544287.1 | 3 - 4 | 1244 | 79 | ||
XM_011545986.2 | XP_011544288.1 | 3 - 4 | 1244 | 79 | ||
GenBank mRNA | AB020688.1 | - | 3 - 4 | 604 | 79 | |
AF061943.1 | - | 3 - 4 | 959 | 79 | ||
AF263313.1 | - | 2 - 3 | 201 | 79 | ||
AK291473.1 | - | 3 - 4 | 535 | 79 | ||
BC015204.1 | - | 3 - 4 | 984 | 79 | ||
BC031825.2 | - | 3 - 4 | 276 | 79 | ||
BC051798.1 | - | 3 - 4 | 754 | 79 | ||
BC054495.1 | - | 3 - 4 | 971 | 79 | ||
BC136653.1 | - | 3 - 4 | 703 | 9020643 | 79 | |
BC136655.1 | - | 3 - 4 | 703 | 9020645 | 79 | |
BC142663.1 | - | 3 - 4 | 358 | 40148112 | 79 | |
BC144344.1 | - | 3 - 4 | 703 | 9052864 | 79 | |
BC151221.1 | - | 3 - 4 | 358 | 40148752 | 79 | |
BC152413.1 | - | 3 - 4 | 604 | 8862518 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |