Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007225.hg.1 |
TC01000467.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007225.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007225.hg.1 |
ENST00000468594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007225.hg.1 |
ENST00000540867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007225.hg.1 |
ENST00000543954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007225.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007791.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154898.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154898.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154898.hg.1 |
ENST00000373178.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154898.hg.1 |
NM_017825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154898.hg.1 |
TC0100007791.hg.1 |
Clariom D TC |