Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000615847.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000615847.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000615847.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000615847.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
ENST00000615847.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006505.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006505.hg.1 |
ENST00000397841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006505.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006505.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006505.hg.1 |
TC21000539.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006504.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006504.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006504.hg.1 |
TC21000539.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000615847.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000615847.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000615847.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000615847.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
ENST00000615847.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008395.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100156978.hg.1 |
TC2100008395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100156978.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100156978.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100156978.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100156978.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100156978.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100156978.hg.1 |
ENST00000397841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100156978.hg.1 |
NR_027128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100156978.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100156976.hg.1 |
TC2100008395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100156976.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100156976.hg.1 |
NR_027129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100156976.hg.1 |
ENST00000330551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100156976.hg.1 |
NR_027129.1 |
Refseq |