Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022424.hg.1 |
ENST00000313683.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022424.hg.1 |
ENST00000438932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022424.hg.1 |
NM_001103149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022424.hg.1 |
NM_018215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022424.hg.1 |
TC19001647.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022426.hg.1 |
ENST00000313683.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022426.hg.1 |
ENST00000438932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022426.hg.1 |
NM_001103149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022426.hg.1 |
NM_018215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022426.hg.1 |
TC19001647.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022425.hg.1 |
ENST00000313683.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022425.hg.1 |
ENST00000417103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022425.hg.1 |
ENST00000438932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022425.hg.1 |
NM_001103149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022425.hg.1 |
NM_018215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022425.hg.1 |
TC19001647.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900191446.hg.1 |
ENST00000313683.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191446.hg.1 |
ENST00000438932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191446.hg.1 |
NM_001103149.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191446.hg.1 |
NM_018215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191446.hg.1 |
TC1900010988.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191446.hg.1 |
ENST00000313683.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191446.hg.1 |
ENST00000438932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191446.hg.1 |
NM_001103149.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191446.hg.1 |
NM_018215.3 |
Refseq |