Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008220.1 | NP_001008221.1 | 4 | 3259 | 70 | |
NM_006650.3 | NP_006641.1 | 5 | 3309 | 70 | ||
XM_005265798.1 | XP_005265855.1 | 4 | 3187 | 70 | ||
XM_005265799.1 | XP_005265856.1 | 4 | 3297 | 70 | ||
XM_011534419.1 | XP_011532721.1 | 4 | 3575 | 70 | ||
XM_017008964.1 | XP_016864453.1 | 4 | 3317 | 70 | ||
GenBank mRNA | AK094266.1 | - | 1 | 1150 | 70 | |
AK124837.1 | - | 2 | 2410 | 70 | ||
AK126590.1 | - | 1 | 598 | 70 | ||
AL512758.1 | - | 1 | 672 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |