Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021455.hg.1 |
ENST00000341918.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021455.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021455.hg.1 |
TC19001574.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021456.hg.1 |
ENST00000341918.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021456.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021456.hg.1 |
TC19001574.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021459.hg.1 |
ENST00000341918.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021459.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021459.hg.1 |
TC19001574.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021457.hg.1 |
ENST00000341918.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021457.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021457.hg.1 |
TC19001574.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900189429.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189429.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189429.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900189429.hg.1 |
TC1900010805.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900189429.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189429.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189429.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010805.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900189428.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189428.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189428.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900189428.hg.1 |
TC1900010805.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900189428.hg.1 |
ENST00000341918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189428.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189428.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900189431.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189431.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900189431.hg.1 |
TC1900010805.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900189431.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189431.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900189433.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189433.hg.1 |
NM_198477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900189433.hg.1 |
TC1900010805.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900189433.hg.1 |
ENST00000601181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189433.hg.1 |
NM_198477.2 |
Refseq |