Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161390.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161390.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161390.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161390.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161390.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161390.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161390.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161390.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161390.hg.1 |
TC1000008974.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000161393.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161393.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161393.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161393.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161393.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161393.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161393.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161393.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161393.hg.1 |
TC1000008974.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000161389.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161389.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161389.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161389.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161389.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161389.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161389.hg.1 |
TC1000008974.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000161388.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161388.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161388.hg.1 |
TC1000008974.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000161392.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161392.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161392.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161392.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161392.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161392.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161392.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161392.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161392.hg.1 |
TC1000008974.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
ENST00000603594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008974.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161394.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161394.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161394.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161394.hg.1 |
ENST00000298715.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161394.hg.1 |
ENST00000392982.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161394.hg.1 |
NM_001272046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161394.hg.1 |
TC1000008974.hg.1 |
Clariom D TC |