Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168074.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168074.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168074.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168074.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168074.hg.1 |
TC0400009894.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400168073.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168073.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168073.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168073.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168073.hg.1 |
TC0400009894.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000324058.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009894.hg.1 |
ENST00000531445.1 |
Ensembl |