Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000412609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000458539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598164.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598164.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598164.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
ENST00000598736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011731.hg.1 |
NR_034156.1 |
Refseq |