Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014000.hg.1 |
ENST00000355025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014000.hg.1 |
ENST00000396829.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014000.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014000.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014000.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014000.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014000.hg.1 |
TC17001040.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173548.hg.1 |
TC1700009544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000355025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000574617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000575780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
ENST00000576820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_001167987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009544.hg.1 |
NM_213726.2 |
Refseq |