Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800145950.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800145950.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800145950.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800145950.hg.1 |
TC1800006720.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800145950.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800145950.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800145950.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000418274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
ENST00000587848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006720.hg.1 |
NM_001277333.1 |
Refseq |