Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018964.hg.1 |
TC01001226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018964.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018964.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018295.hg.1 |
ENST00000434793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018966.hg.1 |
TC01001226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018966.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018966.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018297.hg.1 |
ENST00000615904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018297.hg.1 |
ENST00000615904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018293.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018293.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018293.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018293.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018293.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018293.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018297.hg.1 |
ENST00000615904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018297.hg.1 |
ENST00000615904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100239442.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100239442.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100239442.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100239442.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100239442.hg.1 |
TC0100018293.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018293.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018293.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018293.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018293.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018293.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018293.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100239444.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100239444.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100239444.hg.1 |
ENST00000334269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100239444.hg.1 |
NM_178438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100239444.hg.1 |
TC0100018293.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018295.hg.1 |
ENST00000434793.1 |
Ensembl |