Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000525527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000525527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000525527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000525527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000525527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000525890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000525890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000525890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000515029.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000525527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000525527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000525527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000525527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000525527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000525890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000525890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000525890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000529065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013225.hg.1 |
ENST00000530229.5 |
Ensembl |