Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005602.hg.1 |
ENST00000335099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005602.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005602.hg.1 |
ENST00000533205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005602.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005602.hg.1 |
TC11000464.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005601.hg.1 |
ENST00000335099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005601.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005601.hg.1 |
ENST00000533205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005601.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005601.hg.1 |
TC11000464.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005605.hg.1 |
ENST00000335099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005605.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005605.hg.1 |
TC11000464.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000533205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000533205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
ENST00000533205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153967.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153967.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153967.hg.1 |
ENST00000533205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153967.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153967.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153967.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153967.hg.1 |
ENST00000533205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153967.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153967.hg.1 |
TC1100007675.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100153968.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153968.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153968.hg.1 |
ENST00000533205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153968.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153968.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153968.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153968.hg.1 |
ENST00000533205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153968.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153968.hg.1 |
TC1100007675.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000395120.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000533205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000533205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
ENST00000533205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007675.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153964.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153964.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153964.hg.1 |
ENST00000335099.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153964.hg.1 |
NM_178570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153964.hg.1 |
TC1100007675.hg.1 |
Clariom D TC |