Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000243.hg.1 |
TC0X000015.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000243.hg.1 |
ENST00000381174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000243.hg.1 |
ENST00000419513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000243.hg.1 |
ENST00000426774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000243.hg.1 |
ENST00000533923.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000243.hg.1 |
NM_001141919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000243.hg.1 |
NM_001141920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000243.hg.1 |
NM_175569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000243.hg.1 |
NR_003254.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000241.hg.1 |
TC0X000015.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000241.hg.1 |
ENST00000381174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000241.hg.1 |
ENST00000419513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000241.hg.1 |
ENST00000426774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000241.hg.1 |
ENST00000533923.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000241.hg.1 |
NM_001141919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000241.hg.1 |
NM_001141920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000241.hg.1 |
NM_175569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000241.hg.1 |
NR_003254.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000188.hg.1 |
TC0Y000352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000188.hg.1 |
NR_003254.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000187.hg.1 |
TC0Y000352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000187.hg.1 |
NR_003254.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
ENST00000381174.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
ENST00000419513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
ENST00000503018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
ENST00000509484.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
NM_001141919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
NM_001141920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
NM_175569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
NR_003254.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
ENST00000381174.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
ENST00000419513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
ENST00000503018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
ENST00000509484.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
NM_001141919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
NM_001141920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
NM_175569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
NR_003254.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144469.hg.1 |
TC0X00006480.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
ENST00000381174.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
ENST00000419513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
ENST00000503018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
NM_001141919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
NM_001141920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
NM_175569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
NR_003254.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
ENST00000381174.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
ENST00000419513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
ENST00000503018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
NM_001141919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
NM_001141920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
NM_175569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
NR_003254.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144470.hg.1 |
TC0X00006480.hg.1 |
Clariom D TC |