Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000843.hg.1 |
TC0Y000073.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000843.hg.1 |
ENST00000288666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000843.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00145828.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00145828.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00145828.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00145828.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00145828.hg.1 |
TC0Y00006732.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
ENST00000629237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006732.hg.1 |
NM_001039567.2 |
Refseq |