Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_001124759_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8080995 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7937256 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8080997 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8080996 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8104128 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7937253 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7937255 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8104125 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8104127 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7937251 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8104124 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8080994 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000411596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000411596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000411596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000419104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000419104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000419104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000419104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000419104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000422555.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000422555.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000422555.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000422555.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
ENST00000425520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
ENST00000425520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
ENST00000425520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
ENST00000425520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
ENST00000425520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
NM_001080998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
NM_001080998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
NM_001080998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
NM_001080998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012416.hg.1 |
NM_001080998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156542.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156542.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156542.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156542.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156542.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156542.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156542.hg.1 |
TC0300007883.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300156539.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156539.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156539.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156539.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156539.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156539.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156539.hg.1 |
TC0300007883.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300156544.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156544.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156544.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156544.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156544.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156544.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156544.hg.1 |
TC0300007883.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300156541.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156541.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156541.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156541.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156541.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156541.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156541.hg.1 |
TC0300007883.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
ENST00000425520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
ENST00000425520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
ENST00000425520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
ENST00000425520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
ENST00000425520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
ENST00000443774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
NM_001080998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
NM_001080998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
NM_001080998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
NM_001080998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012416.hg.1 |
NM_001080998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156538.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156538.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156538.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156538.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156538.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156538.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156538.hg.1 |
TC0300007883.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000411596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000411596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000411596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000419104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000419104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000419104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000419104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000419104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000422555.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000422555.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000422555.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000422555.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010078.hg.1 |
ENST00000453982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156543.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156543.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156543.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156543.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156543.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156543.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156543.hg.1 |
TC0300007883.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000308062.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
ENST00000464571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007883.hg.1 |
NM_001124759.3 |
Refseq |