Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009775.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009775.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009775.hg.1 |
TC14000902.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009769.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009769.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009769.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009769.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009769.hg.1 |
ENST00000555698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009769.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009769.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009769.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009769.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009769.hg.1 |
TC14000902.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000555698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000555698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000555698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
ENST00000555698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000555698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000555698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000555698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
ENST00000555698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008606.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
ENST00000555698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
ENST00000340900.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
ENST00000397967.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
ENST00000397970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
ENST00000555698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
NM_002933.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
NM_198234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
NM_198235.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161909.hg.1 |
TC1400008606.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400161915.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161915.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161915.hg.1 |
ENST00000412779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161915.hg.1 |
NM_198232.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161915.hg.1 |
TC1400008606.hg.1 |
Clariom D TC |