Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
7979926 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7979925 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009578.hg.1 |
ENST00000320746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009578.hg.1 |
ENST00000556964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009578.hg.1 |
ENST00000556964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009578.hg.1 |
ENST00000556964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009578.hg.1 |
ENST00000556964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009578.hg.1 |
NR_037933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009578.hg.1 |
NR_037933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009578.hg.1 |
NR_037933.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009578.hg.1 |
NR_037933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400170598.hg.1 |
ENST00000556964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400170598.hg.1 |
NR_037933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400170598.hg.1 |
ENST00000556964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400170598.hg.1 |
NR_037933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400170598.hg.1 |
TC1400009578.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400009578.hg.1 |
ENST00000320746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009578.hg.1 |
ENST00000556964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009578.hg.1 |
ENST00000556964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009578.hg.1 |
ENST00000556964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009578.hg.1 |
ENST00000556964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009578.hg.1 |
NR_037933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009578.hg.1 |
NR_037933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009578.hg.1 |
NR_037933.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009578.hg.1 |
NR_037933.1 |
Refseq |