Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS6ST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153456.3 | NP_703157.2 | 2 | 6869 | 138 | |
GenBank mRNA | AF339796.1 | - | 1 | 559 | 138 | |
AK091343.1 | - | 1 | 1120 | 138 | ||
BC037795.1 | - | 1 | 935 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |