Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007232.1 | NP_001007233.1 | 2 | 180 | 118 | |
XM_011542838.2 | XP_011541140.1 | 2 | 236 | 118 | ||
GenBank mRNA | AY761064.1 | - | 2 | 180 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16498040 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16498056 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428486 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002334 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428449 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428439 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428384 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002338 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428377 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428417 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16498036 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428383 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428385 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428450 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428391 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002310 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428409 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16498012 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428408 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002304 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16498024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002313 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428410 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002327 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002294 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002302 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002349 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16498075 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002320 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002311 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16498009 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428495 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428373 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002359 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428416 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002307 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002303 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428492 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002323 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428411 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428448 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16498014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428378 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428429 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428459 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428488 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16498083 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002330 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002358 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428422 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002305 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428479 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002308 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002321 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428494 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428489 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002343 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28428515 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002361 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08002353 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 519155 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 337283 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 24961 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 40660 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 40918 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 171093 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 357100 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 317472 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 382810 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 92026 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 212950 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 137747 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P192805 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P192805 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 13935 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 58162 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P192805 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P192805 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P192805 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002354 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002349 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002339 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002343 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002342 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002345 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002356 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002341 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002347 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002352 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002340 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002350 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002355 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08002348 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02510851 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P192805 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02510856 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19356438 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02510851 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19356463 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02510847 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002345 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002341 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002316 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002302 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002325 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002322 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002330 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002337 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002328 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002290 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002359 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002310 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002307 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002347 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002352 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002311 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002317 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002336 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002354 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002295 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002334 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002293 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08002315 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02510851 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02510856 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001007232_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7951416 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7951414 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7951415 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7951413 | - | - |
Affymetrix Human Genome U219 Array | 11764103_at | - | - |
Affymetrix Human Genome U219 Array | 11738958_at | - | - |
Affymetrix Human Genome U219 Array | 11764103_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738959_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11738960_x_at | - | - |
Affymetrix Human Genome U219 Array | 11738958_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738959_s_at | - | - |
Affymetrix Human Genome U219 Array | 11738960_PM_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR11026933.hg.1 | TC11003507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11026933.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026933.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013223.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013223.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013223.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013223.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013223.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013223.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012169.hg.1 | ENST00000526345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012169.hg.1 | ENST00000526345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026934.hg.1 | TC11003507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11026934.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026934.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11026935.hg.1 | TC11003507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11026935.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11026935.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100205327.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100205327.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100205327.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100205327.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100205327.hg.1 | TC1100013223.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100012169.hg.1 | ENST00000526345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012169.hg.1 | ENST00000526345.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100205328.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100205328.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100205328.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100205328.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100205328.hg.1 | TC1100013223.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100205326.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100205326.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100205326.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100205326.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100205326.hg.1 | TC1100013223.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100013223.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013223.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013223.hg.1 | ENST00000375707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013223.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013223.hg.1 | NM_001007232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013223.hg.1 | NM_001007232.1 | Refseq |
Human Genome U95 Set | 52066_PM_at | - | - |
Human Genome U95 Set | 52066_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045647 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045647 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 219367 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 222692 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3238525 | NM_001007232.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3238525 | AY761064.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1707979 | AY761064.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1707979 | AY761064.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3238525 | NM_001007232.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3238525 | AY761064.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1707979 | AY761064.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238525 | NM_001007232.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238525 | AY761064.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1707979 | AY761064.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238525 | NM_001007232.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238525 | AY761064.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1707979 | AY761064.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_6860-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1707979 | AY761064.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1707979 | AY761064.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 1470184 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_6860 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1707979 | AY761064.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1707979 | AY761064.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1707979 | AY761064.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1707979 | AY761064.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1707979 | NM_001007232.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1707979 | AY761064.1 | human_transcriptome |