Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRXL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002223.3 | 3 | 1880 | 96 | ||
GenBank mRNA | AK092426.1 | - | 3 | 1805 | 96 | |
BC027729.2 | - | 5 | 1885 | 96 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
FYVE, RhoGEF and PH domain containing 5 pseudogene 1 |
FGD5P1 | -128.3kb | Anti-sense | Chr. 3 - 13933056 - 13936944 on NCBI Build GRCh38 | |
Downstream | coiled-coil-helix-coiled-coil-helix domain containing 4 | CHCHD4 |
MIA40, TIMM40 |
46.1kb | Sense | Chr. 3 - 14112077 - 14124871 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |