Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |