Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002725.3 | NP_002716.1 | 3 | 5328 | 116 | |
NM_201348.1 | NP_958505.1 | 3 | 5318 | 116 | ||
GenBank mRNA | AK098245.1 | - | 1 | 1625 | 116 | |
BQ010158.1 | - | 1 | 116 | |||
BQ182260.1 | - | 2 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |