Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021018.2 | NP_066298.1 | 1 | 351 | 120 | |
GenBank mRNA | BC067492.1 | - | 1 | 349 | 7002073 | 120 |
BC069818.1 | - | 1 | 351 | 7262721 | 120 | |
BC096131.2 | - | 1 | 349 | 40002512 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |