Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023504.hg.1 |
TC04001699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023504.hg.1 |
ENST00000280605.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023504.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023509.hg.1 |
TC04001699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023509.hg.1 |
ENST00000280605.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023509.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023505.hg.1 |
TC04001699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023505.hg.1 |
ENST00000280605.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023505.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023508.hg.1 |
TC04001699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023508.hg.1 |
ENST00000280605.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023508.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184367.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184367.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184367.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184367.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184367.hg.1 |
TC0400012309.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012309.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184372.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184372.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184372.hg.1 |
TC0400012309.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400184366.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184366.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184366.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184366.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184366.hg.1 |
TC0400012309.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400184368.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184368.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184368.hg.1 |
ENST00000280605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184368.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184368.hg.1 |
TC0400012309.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400184373.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184373.hg.1 |
NM_032136.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184373.hg.1 |
TC0400012309.hg.1 |
Clariom D TC |