Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002255.1 | NP_001002255.1 | 1 | 461 | 141 | |
GenBank mRNA | AY340238.1 | - | 1 | 461 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364081 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364109 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364075 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364086 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364104 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05163923 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364094 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364074 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364079 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364102 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364093 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364111 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05163915 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364114 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05163933 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05163938 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364107 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364112 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364106 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364085 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364084 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364089 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05163948 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364092 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15872281 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364083 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364115 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364100 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364113 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364099 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15872290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364090 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05163940 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364108 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364095 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364073 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25364101 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0235 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P118435 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P330691 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364108 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364095 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364112 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364090 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05163940 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364081 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364092 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05163948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364099 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364084 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364111 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364087 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15872281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15872290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364086 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364123 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364106 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364109 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364083 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05163923 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364115 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05163958 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364085 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364100 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364101 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364089 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364093 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364079 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364078 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364086 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05163940 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364087 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364111 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15872281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364078 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364083 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364089 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364100 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05163948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364108 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364112 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364093 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364090 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364084 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364095 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364123 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364081 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364109 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364101 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05163923 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05163958 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364085 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364079 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364106 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364099 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15872290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364115 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25364092 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163935 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163915 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163955 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163936 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163926 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163918 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163950 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163958 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163942 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163920 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163939 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15872280 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15872276 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163949 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163924 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163933 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15872284 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15872275 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163946 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163929 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15872282 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163944 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05163931 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001002255_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8174637 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8122686 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8178323 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8174640 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8122684 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 215452_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11740798_at | - | - |
Affymetrix Human Genome U219 Array | 11740798_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR06012787.hg.1 | TC06001079.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06012787.hg.1 | ENST00000326669.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06012787.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06012790.hg.1 | TC06001079.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06012790.hg.1 | ENST00000326669.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06012790.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009795.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06012789.hg.1 | TC06001079.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06012789.hg.1 | ENST00000326669.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06012789.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167090.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167090.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167090.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167090.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167090.hg.1 | TC0600009795.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009795.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167089.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167089.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167089.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167089.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167089.hg.1 | TC0600009795.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600167088.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167088.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167088.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167088.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167088.hg.1 | TC0600009795.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600167091.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167091.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167091.hg.1 | ENST00000326669.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600167091.hg.1 | NM_001002255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600167091.hg.1 | TC0600009795.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 208738_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 215452_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 208738_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 215452_x_at | - | - |
Human Genome U133A 2.0 Array | 215452_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 215452_x_at | - | - |
Human Genome U133A 2.0 Array | 208738_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 208738_x_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_001002255_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw503128 | - | - |
Human Genome U95 Set | 41186_PM_at | - | - |
Human Genome U95 Set | 32478_PM_f_at | - | - |
Human Genome U95 Set | 32478_f_at | - | - |
Human Genome U95 Set | 41186_at | - | - |
Human Genome U133 Set | 215452_PM_x_at | - | - |
Human Genome U133 Set | 215452_x_at | - | - |
Human Genome U133 Set | 208738_PM_x_at | - | - |
Human Genome U133 Set | 208738_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032191 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032191 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1665203 | AY340238.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1665203 | BC130305.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1665203 | AY340238.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1665203 | BC130305.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1665203 | AY340238.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1665203 | BC130305.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1665203 | AY340238.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1665203 | BC130305.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1665203 | AY340238.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1665203 | BC130305.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_17416-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1665203 | AY340238.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1665203 | BC130305.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1665203 | AY340238.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1665203 | BC130305.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 6060475 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_17416 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1665203 | AY340238.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1665203 | BC130305.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1665203 | AY340238.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1665203 | BC130305.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1665203 | AY340238.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1665203 | BC130305.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1665203 | AY340238.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1665203 | BC130305.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1665203 | NM_001002255.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1665203 | AY340238.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1665203 | BC130305.1 | human_transcriptome |