Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020908.hg.1 |
ENST00000302610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020908.hg.1 |
NM_001005211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020908.hg.1 |
TC11001802.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010925.hg.1 |
ENST00000302610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010925.hg.1 |
NM_001005211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010924.hg.1 |
ENST00000608484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010924.hg.1 |
ENST00000608484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010925.hg.1 |
ENST00000302610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010925.hg.1 |
NM_001005211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182556.hg.1 |
ENST00000302610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182556.hg.1 |
NM_001005211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182556.hg.1 |
ENST00000302610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182556.hg.1 |
NM_001005211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182556.hg.1 |
TC1100010925.hg.1 |
Clariom D TC |