Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008540.1 | NP_001008540.1 | 1 | 363 | 88 | |
GenBank mRNA | AF147204.1 | - | 1 | 401 | 88 | |
AK309641.1 | - | 1 | 291 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon |