Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8079741 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8079740 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
230587_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007369.hg.1 |
ENST00000440528.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007369.hg.1 |
ENST00000622888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007369.hg.1 |
ENST00000440528.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007369.hg.1 |
ENST00000622888.1 |
Ensembl |
Human Genome U133 Plus 2.0 Array |
230587_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
230587_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230587_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230587_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230587_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230587_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230587_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230587_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230587_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230587_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230587_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230587_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
230587_at3 |
- |
- |
Human Genome U95 Set |
53574_at |
- |
- |
Human Genome U95 Set |
53574_PM_at |
- |
- |
Human Genome U133 Set |
230587_PM_at |
- |
- |
Human Genome U133 Set |
230587_at |
- |
- |