Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000485151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
ENST00000485151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022616.hg.1 |
TC01001457.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022616.hg.1 |
ENST00000361200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022616.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022616.hg.1 |
ENST00000485151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022616.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022615.hg.1 |
TC01001457.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022615.hg.1 |
ENST00000361200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022615.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022615.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177195.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177195.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177195.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177195.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177195.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177195.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177195.hg.1 |
TC0100010497.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100177196.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177196.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177196.hg.1 |
ENST00000485151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177196.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177196.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177196.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177196.hg.1 |
ENST00000485151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177196.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177196.hg.1 |
TC0100010497.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000271385.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000361200.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000443333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000485151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
ENST00000485151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010497.hg.1 |
NM_001080426.2 |
Refseq |