Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000470589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000472788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
ENST00000621172.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011802.hg.1 |
NM_001105580.2 |
Refseq |