Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAMEF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100631.1 | NP_001094101.1 | 2 - 3 | 872 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Homo sapiens Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 41645 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 495171 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 258729 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 105877 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 235939 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3313755 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3227876 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1021 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P204690 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1020 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 22951 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22610 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 54459 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P10032798 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3313755 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3313755 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3227876 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3232778 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3313755 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3227876 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P204690 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P204690 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7897998 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898000 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898000 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898004 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912588 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912586 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7897999 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912588 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912587 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912605 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898001 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912620 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912587 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912586 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912620 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912619 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898004 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912604 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912604 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912603 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898003 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912602 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912618 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912603 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912605 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912621 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898003 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7897999 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912619 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898001 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7912621 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7912617 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7912585 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7897997 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7912585 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7912617 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7898002 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7912601 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7897997 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7912601 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7898002 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR01002766.hg.1 | TC01000168.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01002766.hg.1 | ENST00000376187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012942.hg.1 | ENST00000624297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012942.hg.1 | ENST00000624297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012942.hg.1 | ENST00000624297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012942.hg.1 | ENST00000624297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012942.hg.1 | NM_001099850.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012942.hg.1 | NM_001099850.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012942.hg.1 | NM_001099850.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012942.hg.1 | NM_001099850.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012948.hg.1 | ENST00000376101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012948.hg.1 | ENST00000376101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012948.hg.1 | ENST00000376101.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012948.hg.1 | NM_001099790.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012948.hg.1 | NM_001099790.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012948.hg.1 | NM_001099790.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012948.hg.1 | NM_001099790.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012948.hg.1 | NM_001099790.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012948.hg.1 | ENST00000376101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012948.hg.1 | ENST00000376101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012948.hg.1 | ENST00000376101.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012948.hg.1 | NM_001099790.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012948.hg.1 | NM_001099790.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012948.hg.1 | NM_001099790.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012948.hg.1 | NM_001099790.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012948.hg.1 | NM_001099790.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012942.hg.1 | ENST00000624297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012942.hg.1 | ENST00000624297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012942.hg.1 | ENST00000624297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012942.hg.1 | ENST00000624297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012942.hg.1 | NM_001099850.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012942.hg.1 | NM_001099850.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012942.hg.1 | NM_001099850.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012942.hg.1 | NM_001099850.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016148 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011361 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011583 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016148 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016148 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011583 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016148 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011361 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3236604 | NM_001100631.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3300849 | NM_001099790.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3300849 | NM_001013692.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3300849 | NM_001099850.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3227049 | NM_001099790.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3227049 | NM_001013692.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3227049 | NM_001099850.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1771291 | NM_001013692.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3300849 | NM_001099790.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3300849 | NM_001013692.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3300849 | NM_001099850.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3236604 | NM_001100631.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3300849 | NM_001099790.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3300849 | NM_001013692.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3300849 | NM_001099850.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236604 | NM_001100631.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236604 | NM_001100631.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3300849 | NM_001099790.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3300849 | NM_001013692.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3300849 | NM_001099850.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_27018-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1771291 | NM_001013692.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1771291 | NM_001013692.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 7550563 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_27018 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1771291 | NM_001013692.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1771291 | NM_001013692.1 | human_transcriptome |