Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000297574.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000297574.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000297574.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000297574.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000297574.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000297574.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000297574.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000297574.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000306391.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000306391.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000309982.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000309982.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000309982.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000309982.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000309982.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000309982.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000309982.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000309982.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000309982.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000309982.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000309982.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000330955.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000330955.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000438105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000438105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000438105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000438105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000438105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000438105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000438105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000438105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000438105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000438105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000517863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000517863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000517863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000519507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000519507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000519507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000519507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000521926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000521926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000521926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000521926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000521926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000523754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000523754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000523754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000523754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000523754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000523754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000523754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
ENST00000523754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_001024372.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_001024372.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_001024372.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_001024372.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_001024372.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_001024372.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_001024372.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_001024372.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_001024372.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_001024372.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_024812.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_024812.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_024812.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_024812.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_024812.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_024812.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_024812.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_024812.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_024812.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_024812.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008474.hg.1 |
NM_024812.2 |
Refseq |