Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000631833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
ENST00000632230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011305.hg.1 |
NM_001013627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177671.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177671.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177671.hg.1 |
ENST00000373677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177671.hg.1 |
ENST00000510661.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177671.hg.1 |
TC0X00011305.hg.1 |
Clariom D TC |