Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003075.3 | NP_003066.2 | 27 | 3386 | 129 | |
XM_005269101.1 | XP_005269158.1 | 28 | 3416 | 129 | ||
XM_005269102.1 | XP_005269159.1 | 28 | 3413 | 129 | ||
XM_005269103.1 | XP_005269160.1 | 27 | 3320 | 129 | ||
XM_011538693.2 | XP_011536995.1 | 27 | 3325 | 129 | ||
XM_017019887.1 | XP_016875376.1 | 26 | 3210 | 129 | ||
GenBank mRNA | AK315153.1 | - | 27 | 3283 | 129 | |
BC009067.1 | - | 6 | 1034 | 129 | ||
BC026222.2 | - | 27 | 3305 | 4395012 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |