Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003481.hg.1 |
ENST00000268379.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003481.hg.1 |
NM_003366.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003481.hg.1 |
TC16000239.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
ENST00000268379.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
ENST00000561553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
ENST00000563711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
ENST00000563898.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
ENST00000567810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
NM_003366.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
ENST00000268379.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
ENST00000561553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
ENST00000563711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
ENST00000563898.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
ENST00000567810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
NM_003366.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600151427.hg.1 |
TC1600007183.hg.1 |
Clariom D TC |