Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173808.2 | NP_776169.2 | 7 | 2254 | 72 | |
GenBank mRNA | AK092307.1 | - | 7 | 2161 | 72 | |
BC036771.1 | - | 7 | 2176 | 5576828 | 72 | |
BX538014.1 | - | 7 | 2158 | 72 | ||
BX648027.1 | - | 3 | 1385 | 72 | ||
CR749215.1 | - | 2 | 1338 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |