Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612817.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612817.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612817.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000612817.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000617461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000617461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000617461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000617461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010271.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010271.hg.1 |
TC22000581.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
TC2200008188.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158663.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000611600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612817.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612817.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612817.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000612817.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000617461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000617461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000617461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000617461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000617532.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
ENST00000621179.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008188.hg.1 |
NR_003082.1 |
Refseq |