Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019864.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019864.hg.1 |
TC12001510.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019865.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019865.hg.1 |
TC12001510.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010713.hg.1 |
ENST00000564840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010713.hg.1 |
ENST00000564840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010713.hg.1 |
ENST00000618634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010713.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010713.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010713.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010713.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010713.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181723.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181723.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181723.hg.1 |
TC1200010713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200181721.hg.1 |
ENST00000564840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181721.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181721.hg.1 |
ENST00000564840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181721.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181721.hg.1 |
TC1200010713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200181720.hg.1 |
ENST00000564840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181720.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181720.hg.1 |
ENST00000564840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181720.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181720.hg.1 |
TC1200010713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010713.hg.1 |
ENST00000564840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010713.hg.1 |
ENST00000564840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010713.hg.1 |
ENST00000618634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010713.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010713.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010713.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010713.hg.1 |
NM_001013690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010713.hg.1 |
NM_001013690.4 |
Refseq |